Product Details

SNP ID
rs972100653
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:115267281 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGAAGGAAACCAAAAAGATTATAT[C/T]AATAATACAATTAATAGGGTATATG
Phenotype
MIM: 602031
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCDC112 PubMed Links

Gene Details

Gene
CCDC112
Gene Name
coiled-coil domain containing 112
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040440.2 2449 UTR 3 NP_001035530.1
NM_152549.2 2449 UTR 3 NP_689762.2
Gene
PGGT1B
Gene Name
protein geranylgeranyltransferase type I subunit beta
There are no transcripts associated with this gene.

View Full Product Details