Product Details

SNP ID
rs121434367
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:12899486 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTCCAGGTACACATGACATTCACG[C/T]CCTGATCCTTGGGAGAGCTATCACG
Phenotype
MIM: 608801 MIM: 611487
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GCDH PubMed Links

Gene Details

Gene
GCDH
Gene Name
glutaryl-CoA dehydrogenase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000159.3 1370 Missense Mutation GCC,GTC A421V NP_000150.1
NM_013976.3 1370 Intron NP_039663.1
XM_006722721.2 1370 Intron XP_006722784.1
XM_011527899.2 1370 Intron XP_011526201.1
XM_011527900.2 1370 Intron XP_011526202.1
XM_017026580.1 1370 Intron XP_016882069.1
Gene
SYCE2
Gene Name
synaptonemal complex central element protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001105578.1 1370 Intron NP_001099048.1
XM_005259848.4 1370 Intron XP_005259905.1
XM_011527882.2 1370 Intron XP_011526184.1
XM_011527883.2 1370 Intron XP_011526185.1

View Full Product Details