Product Details
- SNP ID
-
rs4981823
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:20197728 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCCTCTCTGACACCAAGATCATCTC[C/G]TTCTCTGGCTGCTTCCTCCAGTTCT
- Phenotype
-
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
OR11G2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs77418176] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- OR11G2
- Gene Name
- olfactory receptor family 11 subfamily G member 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001005503.1 |
393 |
Silent Mutation |
TCC,TCG |
S131S |
NP_001005503.1 |
View Full Product Details