Product Details

SNP ID
rs4313359
Assay Type
Functionally tested
NCBI dbSNP Submissions
44
Location
Chr.1:33328081 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCTTGAGAAAGAGGCTTGTGGTT[A/G]CCATGAATACCTGACCTGGTCGGTG
Phenotype
MIM: 602979
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
A3GALT2 PubMed Links

Gene Details

Gene
A3GALT2
Gene Name
alpha 1,3-galactosyltransferase 2
There are no transcripts associated with this gene.

Gene
MIR3605
Gene Name
microRNA 3605
There are no transcripts associated with this gene.

Gene
PHC2
Gene Name
polyhomeotic homolog 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004427.3 Intron NP_004418.2
NM_198040.2 Intron NP_932157.1
XM_005270570.1 Intron XP_005270627.1
XM_011540876.2 Intron XP_011539178.1
XM_011540877.2 Intron XP_011539179.1
XM_011540878.2 Intron XP_011539180.1
XM_017000513.1 Intron XP_016856002.1
XM_017000514.1 Intron XP_016856003.1
XM_017000515.1 Intron XP_016856004.1
XM_017000516.1 Intron XP_016856005.1
XM_017000517.1 Intron XP_016856006.1
XM_017000518.1 Intron XP_016856007.1
XM_017000519.1 Intron XP_016856008.1
XM_017000520.1 Intron XP_016856009.1
XM_017000521.1 Intron XP_016856010.1
XM_017000522.1 Intron XP_016856011.1

View Full Product Details