Product Details

SNP ID
rs4851137
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:98445405 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATGCGGCTCCAGGGCTGGGTCTAC[C/G]GGCTCCTGAAGCTGGAGGCCTTGCC
Phenotype
MIM: 600916
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
INPP4A PubMed Links

Gene Details

Gene
INPP4A
Gene Name
inositol polyphosphate-4-phosphatase type I A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134224.1 Intron NP_001127696.1
NM_001134225.1 Intron NP_001127697.1
NM_001566.2 Intron NP_001557.1
NM_004027.2 Intron NP_004018.1

View Full Product Details