Product Details

SNP ID
rs4900433
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:99513618 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCAGCTAGGGAGAGGGACAGGGCG[A/G]CCATCTCCCTCCTCTTGCCTGCATC
Phenotype
MIM: 603544
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCDC85C PubMed Links

Gene Details

Gene
CCDC85C
Gene Name
coiled-coil domain containing 85C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144995.1 3319 UTR 3 NP_001138467.1
Gene
CCNK
Gene Name
cyclin K
There are no transcripts associated with this gene.

View Full Product Details