Product Details

SNP ID
rs4815837
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:5545002 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGGTGGGTGGTGTGCCTCACATTTA[C/T]AGCCGATCATTTACAGCACATAATT
Phenotype
MIM: 614124
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GPCPD1 PubMed Links

Gene Details

Gene
GPCPD1
Gene Name
glycerophosphocholine phosphodiesterase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019593.3 5042 UTR 3 NP_062539.1
XM_005260758.4 5042 UTR 3 XP_005260815.1
XM_005260761.1 5042 UTR 3 XP_005260818.1
XM_006723596.3 5042 UTR 3 XP_006723659.1

View Full Product Details