Product Details

SNP ID
rs4715651
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:57319578 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GATTGAAAAGTGTCCCTTGGGTTTG[A/G]CACTTGAGGTGAAATGATAAGGGCC
Phenotype
MIM: 176636
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PRIM2 PubMed Links

Gene Details

Gene
PRIM2
Gene Name
primase (DNA) subunit 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000947.4 Intron NP_000938.2
NM_001282487.1 Intron NP_001269416.1
NM_001282488.1 Intron NP_001269417.1
XM_011514731.2 Intron XP_011513033.1
XM_017011004.1 Intron XP_016866493.1
XM_017011005.1 Intron XP_016866494.1
XM_017011006.1 Intron XP_016866495.1
XM_017011007.1 Intron XP_016866496.1

View Full Product Details