Product Details

SNP ID
rs4242647
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:38393212 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCCGCTGTGGGTGAACTGTAAAAAA[C/T]TGCCTGTATTCAGGAGGATATGCAA
Phenotype
MIM: 100670
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ALDH1B1 PubMed Links

Gene Details

Gene
ALDH1B1
Gene Name
aldehyde dehydrogenase 1 family member B1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000692.4 Intron NP_000683.3
XM_011517802.2 Intron XP_011516104.1

View Full Product Details