Product Details
- SNP ID
-
rs4622780
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:99015297 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TATCAAAAAGATAATACACCATGAC[C/G]AAGTGAGTTGCATACCAGGTATGCA
- Phenotype
-
MIM: 607166
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
TSGA10
PubMed Links
Gene Details
- Gene
- TSGA10
- Gene Name
- testis specific 10
View Full Product Details