Product Details

SNP ID
rs5021392
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:70038597 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTAACTAAATACCTAATGAATACAA[C/G]TACAGCAATTCTTAGGAAGCAGAAG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MIR1972-2 PubMed Links

Gene Details

Gene
MIR1972-2
Gene Name
microRNA 1972-2
There are no transcripts associated with this gene.

Gene
PDXDC2P
Gene Name
pyridoxal dependent decarboxylase domain containing 2, pseudogene
There are no transcripts associated with this gene.

View Full Product Details