Product Details

SNP ID
rs2346586
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:199760858 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCACTGGGAATAAGGGAAGGAGAGA[C/T]TAGTATTTGGAAGAGAAAGAGTGGT
Phenotype
MIM: 614308
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FTCDNL1 PubMed Links

Gene Details

Gene
FTCDNL1
Gene Name
formiminotransferase cyclodeaminase N-terminal like
There are no transcripts associated with this gene.

View Full Product Details