Product Details

SNP ID
rs7424166
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:24946287 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGATTCCTCCCAACAGAGGCCCTG[A/G]GATCTCCCCTGACTGCCACCCAAAG
Phenotype
MIM: 613527
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DNAJC27 PubMed Links

Gene Details

Gene
DNAJC27
Gene Name
DnaJ heat shock protein family (Hsp40) member C27
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001198559.1 2232 UTR 3 NP_001185488.1
NM_016544.2 2232 UTR 3 NP_057628.1
XM_011532902.1 2232 Intron XP_011531204.1
XM_017004269.1 2232 UTR 3 XP_016859758.1
XM_017004270.1 2232 UTR 3 XP_016859759.1

View Full Product Details