Product Details

SNP ID
rs7653045
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:46222821 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCAATAATTAAATATGATTCAATA[A/T]GTGTCTGGCACTTGGTAAATGTTCA
Phenotype
MIM: 601268
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CCR3 PubMed Links

Gene Details

Gene
CCR3
Gene Name
C-C motif chemokine receptor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164680.1 Intron NP_001158152.1
NM_001837.3 Intron NP_001828.1
NM_178328.1 Intron NP_847898.1
NM_178329.2 Intron NP_847899.1
XM_006712960.3 Intron XP_006713023.1
XM_011533335.2 Intron XP_011531637.1
XM_017005685.1 Intron XP_016861174.1
XM_017005686.1 Intron XP_016861175.1

View Full Product Details