Product Details

SNP ID
rs7282383
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.21:41800145 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCATTGTAAAGAGCCAAACTCATT[A/C]AATATTTTTATAGAAAACTGGTCAT
Phenotype
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
PRDM15 PubMed Links

Gene Details

Gene
PRDM15
Gene Name
PR domain 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040424.2 4803 UTR 3 NP_001035514.1
NM_001282934.1 4803 UTR 3 NP_001269863.1
NM_022115.4 4803 Intron NP_071398.3
XM_006724039.2 4803 UTR 3 XP_006724102.1
XM_006724040.2 4803 UTR 3 XP_006724103.1
XM_011529674.1 4803 UTR 3 XP_011527976.1
XM_011529675.1 4803 UTR 3 XP_011527977.1
XM_011529676.2 4803 UTR 3 XP_011527978.1
XM_011529677.2 4803 UTR 3 XP_011527979.1
XM_011529678.2 4803 UTR 3 XP_011527980.1
XM_011529679.2 4803 UTR 3 XP_011527981.1
XM_011529681.2 4803 UTR 3 XP_011527983.1
XM_011529683.1 4803 UTR 3 XP_011527985.1
XM_017028425.1 4803 UTR 3 XP_016883914.1
XM_017028426.1 4803 UTR 3 XP_016883915.1

View Full Product Details