Product Details

SNP ID
rs6449959
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:68221735 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGGTTACACAGAAAATCTACTTTA[C/T]AGCAGTTGTACGTTAAGTAGCTGTG
Phenotype
MIM: 171833
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PIK3R1 PubMed Links

Gene Details

Gene
PIK3R1
Gene Name
phosphoinositide-3-kinase regulatory subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242466.1 Intron NP_001229395.1
NM_181504.3 Intron NP_852556.2
NM_181523.2 Intron NP_852664.1
NM_181524.1 Intron NP_852665.1
XM_005248542.3 Intron XP_005248599.1
XM_011543493.2 Intron XP_011541795.1
XM_017009585.1 Intron XP_016865074.1
XM_017009586.1 Intron XP_016865075.1

View Full Product Details