Product Details

SNP ID
rs6549843
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:67241054 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAACGTAGAAATCATTACTGGTCTT[C/T]AGAAAGATGCAAGTTTGCAAACCAG
Phenotype
MIM: 603922
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SUCLG2 PubMed Links

Gene Details

Gene
SUCLG2
Gene Name
succinate-CoA ligase GDP-forming beta subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001177599.1 2081 Intron NP_001171070.1
NM_003848.3 2081 Intron NP_003839.2
XM_017007415.1 2081 Intron XP_016862904.1
XM_017007416.1 2081 Intron XP_016862905.1
XM_017007417.1 2081 Intron XP_016862906.1
XM_017007418.1 2081 Intron XP_016862907.1
XM_017007419.1 2081 UTR 3 XP_016862908.1
XM_017007420.1 2081 Intron XP_016862909.1

View Full Product Details