Product Details
- SNP ID
-
rs7499304
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:20553068 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CATCTTTTAGAAACTGCCTTTTACT[G/T]CAGCTCTTCTATTGTATAATTTTTA
- Phenotype
-
MIM: 614359
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
ACSM2B
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs77260326] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ACSM2B
- Gene Name
- acyl-CoA synthetase medium-chain family member 2B
View Full Product Details