Product Details
- SNP ID
-
rs7043171
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:70386402 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AAGATACAGTGATGTGATTTCATTC[C/G]TAGTATTCCATGTTGCCTGCATTCT
- Phenotype
-
MIM: 602902
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
KLF9
PubMed Links
Gene Details
- Gene
- KLF9
- Gene Name
- Kruppel like factor 9
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001206.2 |
3403 |
UTR 3 |
|
|
NP_001197.1 |
View Full Product Details