Product Details

SNP ID
rs4937114
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:126282721 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTCTCATTTGTATAAAAGGAAAAAG[G/T]TATGAACACCCCCCTTGCAGGGTGT
Phenotype
MIM: 613622 MIM: 606252
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FOXRED1 PubMed Links

Gene Details

Gene
FOXRED1
Gene Name
FAD dependent oxidoreductase domain containing 1
There are no transcripts associated with this gene.

Gene
TIRAP
Gene Name
TIR domain containing adaptor protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039661.1 1439 Intron NP_001034750.1
NM_001318776.1 1439 Intron NP_001305705.1
NM_001318777.1 1439 Intron NP_001305706.1
NM_148910.2 1439 Intron NP_683708.1
XM_005271399.3 1439 Intron XP_005271456.2
XM_011542576.2 1439 Intron XP_011540878.1
XM_017017161.1 1439 UTR 5 XP_016872650.1
XM_017017162.1 1439 Intron XP_016872651.1

View Full Product Details