Product Details

SNP ID
rs7836723
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:144289233 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATCTTCCAGGCCTGAGAACACACT[C/T]TCCTCGCTGTCGGAGACGCCAGAAT
Phenotype
MIM: 610596 MIM: 140580
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BOP1 PubMed Links

Gene Details

Gene
BOP1
Gene Name
block of proliferation 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015201.4 239 Silent Mutation GAA,GAG E57E NP_056016.1
Gene
HSF1
Gene Name
heat shock transcription factor 1
There are no transcripts associated with this gene.

View Full Product Details