Product Details

SNP ID
rs7893169
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:43555267 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCTGTAGGATTCCCGAATTCCCATA[A/G]TACAGAGGTCTTTCCCTTGGGGCCA
Phenotype
MIM: 601069
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF239 PubMed Links

Gene Details

Gene
ZNF239
Gene Name
zinc finger protein 239
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001099282.1 3070 Intron NP_001092752.1
NM_001099283.1 3070 Intron NP_001092753.1
NM_001099284.1 3070 Intron NP_001092754.1
NM_001324347.1 3070 Intron NP_001311276.1
NM_001324348.1 3070 Intron NP_001311277.1
NM_001324349.1 3070 Intron NP_001311278.1
NM_001324350.1 3070 Intron NP_001311279.1
NM_001324351.1 3070 Intron NP_001311280.1
NM_001324352.1 3070 Intron NP_001311281.1
NM_001324353.1 3070 Intron NP_001311282.1
NM_005674.2 3070 Intron NP_005665.2
XM_005271832.2 3070 UTR 3 XP_005271889.1
XM_006718001.2 3070 UTR 3 XP_006718064.1
XM_006718003.3 3070 UTR 3 XP_006718066.1
XM_011540232.2 3070 UTR 3 XP_011538534.1
XM_011540234.2 3070 UTR 3 XP_011538536.1
XM_011540235.2 3070 UTR 3 XP_011538537.1
XM_011540236.2 3070 UTR 3 XP_011538538.1
XM_011540237.2 3070 UTR 3 XP_011538539.1
XM_011540238.2 3070 UTR 3 XP_011538540.1
XM_017016740.1 3070 UTR 3 XP_016872229.1

View Full Product Details