Product Details

SNP ID
rs6667509
Assay Type
Functionally tested
NCBI dbSNP Submissions
46
Location
Chr.1:215623008 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAATTAAAAGTTCCTTATTAATGAA[G/T]TCTTCCACTTGGACTATGTTTTATT
Phenotype
MIM: 613272 MIM: 608400
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
KCTD3 PubMed Links

Gene Details

Gene
KCTD3
Gene Name
potassium channel tetramerization domain containing 3
There are no transcripts associated with this gene.

Gene
USH2A
Gene Name
usherin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007123.5 18769 Intron NP_009054.5
NM_206933.2 18769 UTR 3 NP_996816.2

View Full Product Details