Product Details
- SNP ID
-
rs7853247
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:87773355 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CAACTTGCAGTTCATAGTATTCAGA[A/T]AGATGAGCTGTTGTCAAAGTTTTGC
- Phenotype
-
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CTSL3P
PubMed Links
Gene Details
- Gene
- CTSL3P
- Gene Name
- cathepsin L family member 3, pseudogene
There are no transcripts associated with this gene.
View Full Product Details