Product Details

SNP ID
rs7150284
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:23377481 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CACCCTTCAGTAGCCTCCTTCCCCA[A/G]GCCCTCTGGACACCTCTCCTGCCCG
Phenotype
MIM: 607888 MIM: 605658 MIM: 160710
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CMTM5 PubMed Links

Gene Details

Gene
CMTM5
Gene Name
CKLF like MARVEL transmembrane domain containing 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001037288.1 Intron NP_001032365.1
NM_001288744.1 Intron NP_001275673.1
NM_001288745.1 Intron NP_001275674.1
NM_001288746.1 Intron NP_001275675.1
NM_138460.2 Intron NP_612469.1
XM_017020953.1 Intron XP_016876442.1
XM_017020954.1 Intron XP_016876443.1
Gene
IL25
Gene Name
interleukin 25
There are no transcripts associated with this gene.

Gene
MYH6
Gene Name
myosin heavy chain 6
There are no transcripts associated with this gene.

View Full Product Details