Product Details

SNP ID
rs6120292
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:31444949 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATTGCTAGGGGGGCATCCCTAGAC[A/T]CCCTATTTATACTGTTAAAATCTGA
Phenotype
MIM: 616076
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
DEFB123 PubMed Links

Gene Details

Gene
DEFB123
Gene Name
defensin beta 123
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153324.3 Intron NP_697019.1

View Full Product Details