Product Details

SNP ID
rs7272371
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:32114132 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCTGTGAACATTCAGGTGTAAGTTC[C/T]TGTGTGAACATACATTCAGTTTCCA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TM9SF4 PubMed Links

Gene Details

Gene
TM9SF4
Gene Name
transmembrane 9 superfamily member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014742.3 Intron NP_055557.2
XM_005260622.4 Intron XP_005260679.1
XM_017028154.1 Intron XP_016883643.1
XM_017028155.1 Intron XP_016883644.1
XM_017028156.1 Intron XP_016883645.1
XM_017028157.1 Intron XP_016883646.1
XM_017028158.1 Intron XP_016883647.1
XM_017028159.1 Intron XP_016883648.1
XM_017028160.1 Intron XP_016883649.1
XM_017028161.1 Intron XP_016883650.1

View Full Product Details