Product Details

SNP ID
rs7855939
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:124456558 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGATCATGGAGCATTTATCCCAGC[A/G]CTCAGGGCAGGCGGCAGTGGGGTGC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ADGRD2 PubMed Links

Gene Details

Gene
ADGRD2
Gene Name
adhesion G protein-coupled receptor D2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_006717348.3 Intron XP_006717411.2

View Full Product Details