Product Details
- SNP ID
-
rs6456144
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:167002117 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AATAGTTAATTTATGCAGCTGTGAA[A/G]TGATCATCACAATCAAGGTACTGCG
- Phenotype
-
MIM: 605392
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
FGFR1OP
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs79378778] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FGFR1OP
- Gene Name
- FGFR1 oncogene partner
- Gene
- MIR3939
- Gene Name
- microRNA 3939
There are no transcripts associated with this gene.
View Full Product Details