Product Details
- SNP ID
-
rs10159323
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
52
- Location
-
Chr.1:227581407 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AGAAAGAAAGGAAAAATTAAAGGAA[A/G]CATGGATTACTAGAAATCTAGATGC
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF678
PubMed Links
Gene Details
- Gene
- ZNF678
- Gene Name
- zinc finger protein 678
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_178549.3 |
|
Intron |
|
|
NP_848644.2 |
View Full Product Details