Product Details

SNP ID
rs10420428
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:44287082 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTAACCAAGTCTAAAACACAGCA[C/T]TTGAGAGACTTCTTTCCTGTCAAGA
Phenotype
MIM: 604749
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF235 PubMed Links

Gene Details

Gene
ZNF235
Gene Name
zinc finger protein 235
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004234.4 6512 UTR 3 NP_004225.3
XM_005259412.4 6512 UTR 3 XP_005259469.1

View Full Product Details