Product Details

SNP ID
rs3110172
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:44752835 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGTTATAATTAGTCAACTTTATAAC[A/G]ATCTAGGGTGAATTAATGCCAACTT
Phenotype
MIM: 616017
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TRIM69 PubMed Links

Gene Details

Gene
TRIM69
Gene Name
tripartite motif containing 69
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301144.1 Intron NP_001288073.1
NM_001301145.1 Intron NP_001288074.1
NM_001301146.1 Intron NP_001288075.1
NM_080745.4 Intron NP_542783.2
NM_182985.4 Intron NP_892030.3

View Full Product Details