Product Details

SNP ID
rs5909309
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:19540527 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAAAAGAACTTAAGTTTGCTTAACC[C/G]AGTTATCCCCCAAAAGTGACCACAA
Phenotype
MIM: 300374
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SH3KBP1 PubMed Links

Gene Details

Gene
SH3KBP1
Gene Name
SH3 domain containing kinase binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001024666.2 Intron NP_001019837.1
NM_001184960.1 Intron NP_001171889.1
NM_031892.2 Intron NP_114098.1
XM_005274494.2 Intron XP_005274551.1
XM_011545498.2 Intron XP_011543800.1
XM_011545499.2 Intron XP_011543801.1
XM_011545500.2 Intron XP_011543802.1
XM_011545502.2 Intron XP_011543804.1
XM_011545503.2 Intron XP_011543805.1
XM_017029459.1 Intron XP_016884948.1
XM_017029460.1 Intron XP_016884949.1
XM_017029461.1 Intron XP_016884950.1
XM_017029462.1 Intron XP_016884951.1
XM_017029463.1 Intron XP_016884952.1
XM_017029464.1 Intron XP_016884953.1
XM_017029465.1 Intron XP_016884954.1
XM_017029466.1 Intron XP_016884955.1
XM_017029467.1 Intron XP_016884956.1
XM_017029468.1 Intron XP_016884957.1
XM_017029469.1 Intron XP_016884958.1
XM_017029470.1 Intron XP_016884959.1

View Full Product Details