Product Details

SNP ID
rs7520495
Assay Type
Functionally tested
NCBI dbSNP Submissions
34
Location
Chr.1:35731409 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTCATGAGAAAGAAGAGTAGACAG[C/G]AGGTTGGGAGGCAGAGCTATGTCAT
Phenotype
MIM: 605434
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CLSPN PubMed Links

Gene Details

Gene
CLSPN
Gene Name
claspin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190481.1 Intron NP_001177410.1
NM_022111.3 Intron NP_071394.2
XM_011541938.2 Intron XP_011540240.1
XM_011541939.2 Intron XP_011540241.1
XM_011541940.2 Intron XP_011540242.1
XM_011541941.2 Intron XP_011540243.1

View Full Product Details