Product Details

SNP ID
rs5915120
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:54442635 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGGGAGACTTTCTTGAGTTACTG[G/T]CTAATTCAGAAAAAGTAGAGCATAG
Phenotype
MIM: 300546 MIM: 300945
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FGD1 PubMed Links

Gene Details

Gene
FGD1
Gene Name
FYVE, RhoGEF and PH domain containing 1
There are no transcripts associated with this gene.

Gene
TSR2
Gene Name
TSR2, ribosome maturation factor
There are no transcripts associated with this gene.

View Full Product Details