Product Details

SNP ID
rs9468791
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:30601549 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGAACAGGGCAGGCAAATGGTCC[C/T]TAGGGCAGGGGGGGCCCATTGACAC
Phenotype
MIM: 603771
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PPP1R10 PubMed Links

Gene Details

Gene
PPP1R10
Gene Name
protein phosphatase 1 regulatory subunit 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002714.3 3429 Silent Mutation TAA,TAG *941* NP_002705.2
XM_006715130.2 3429 Silent Mutation TAA,TAG *941* XP_006715193.1
XM_011514722.1 3429 Silent Mutation TAA,TAG *941* XP_011513024.1
XM_017010995.1 3429 Silent Mutation TAA,TAG *941* XP_016866484.1

View Full Product Details