Product Details

SNP ID
rs10195455
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:192753118 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTATCATAAAAATTATTTGGTTTTA[C/T]CATTTTGACTGTGAGCAAAATATAC
Phenotype
MIM: 605443
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PCGEM1 PubMed Links

Gene Details

Gene
PCGEM1
Gene Name
PCGEM1, prostate-specific transcript (non-protein coding)
There are no transcripts associated with this gene.

View Full Product Details