Product Details
- SNP ID
-
rs6060966
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:31820409 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTGAGACCAGCGTCAAGAAGCCCAA[G/T]GCTGAGCAGGGAGCCTCAGGCAGCC
- Phenotype
-
MIM: 606566
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
MYLK2
PubMed Links
Gene Details
- Gene
- MYLK2
- Gene Name
- myosin light chain kinase 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_033118.3 |
438 |
Missense Mutation |
AAG,AAT |
K112N |
NP_149109.1 |
View Full Product Details