Product Details

SNP ID
rs9962217
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:79970282 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGGGCAAGGTAGACAGCAGGTAC[A/G]TAATGAGATAAACTTGAAATACAGA
Phenotype
MIM: 611595
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HSBP1L1 PubMed Links

Gene Details

Gene
HSBP1L1
Gene Name
heat shock factor binding protein 1 like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136180.1 Intron NP_001129652.1
Gene
TXNL4A
Gene Name
thioredoxin like 4A
There are no transcripts associated with this gene.

View Full Product Details