Product Details

SNP ID
rs6034588
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:16733706 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGATGAGAACAGGCAGTGTTGGCTA[C/T]AGACAGTAGTTTGAGGACCCCTGTT
Phenotype
MIM: 603520
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SNRPB2 PubMed Links

Gene Details

Gene
SNRPB2
Gene Name
small nuclear ribonucleoprotein polypeptide B2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003092.4 Intron NP_003083.1
NM_198220.2 Intron NP_937863.1

View Full Product Details