Product Details

SNP ID
rs10236569
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:128957533 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAAGCTTGGTGGTCTTCAAAGGGTG[G/T]ACTCTGGACACAAACTGCCCGAGTT
Phenotype
MIM: 607218 MIM: 610032
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
IRF5 PubMed Links

Gene Details

Gene
IRF5
Gene Name
interferon regulatory factor 5
There are no transcripts associated with this gene.

Gene
TNPO3
Gene Name
transportin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001191028.2 Intron NP_001177957.2
NM_012470.3 Intron NP_036602.1
XM_011515989.2 Intron XP_011514291.1

View Full Product Details