Product Details

SNP ID
rs10146650
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:45092287 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AACTAACAGTGTCTGCCAGATGACT[A/G]GTGCAACTTTTCCAGAGAGAAATTG
Phenotype
MIM: 614907
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC105370474 PubMed Links

Gene Details

Gene
LOC105370474
Gene Name
uncharacterized LOC105370474
There are no transcripts associated with this gene.

Gene
PRPF39
Gene Name
pre-mRNA processing factor 39
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017922.3 Intron NP_060392.3

View Full Product Details