Product Details

SNP ID
rs10481075
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:124494028 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAAATTTTTTATGACCATTATCTA[A/G]TATATAACCTCTAAATGATAAACCG
Phenotype
MIM: 603046
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RNF139 PubMed Links

Gene Details

Gene
RNF139
Gene Name
ring finger protein 139
There are no transcripts associated with this gene.

Gene
TATDN1
Gene Name
TatD DNase domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146160.1 Intron NP_001139632.1
NM_001317889.1 Intron NP_001304818.1
NM_001317890.1 Intron NP_001304819.1
NM_001317891.1 Intron NP_001304820.1
NM_032026.3 Intron NP_114415.1
XM_006716666.3 Intron XP_006716729.1
XM_006716667.3 Intron XP_006716730.1
XM_006716668.3 Intron XP_006716731.1
XM_006716669.3 Intron XP_006716732.1
XM_006716670.3 Intron XP_006716733.1
XM_006716671.3 Intron XP_006716734.1
XM_011517331.2 Intron XP_011515633.1
XM_017013895.1 Intron XP_016869384.1
XM_017013896.1 Intron XP_016869385.1
XM_017013897.1 Intron XP_016869386.1
XM_017013898.1 Intron XP_016869387.1
XM_017013899.1 Intron XP_016869388.1

View Full Product Details