Product Details

SNP ID
rs10420019
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:22069103 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGCTGGTACTTGGGAGGAAAAGAAG[A/T]AGTTCTGGAGATTCGAACAGTTAAA
Phenotype
MIM: 606957
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ZNF257 PubMed Links

Gene Details

Gene
ZNF257
Gene Name
zinc finger protein 257
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001316996.1 Intron NP_001303925.1
NM_001316997.1 Intron NP_001303926.1
NM_001316998.1 Intron NP_001303927.1
NM_033468.3 Intron NP_258429.2

View Full Product Details