Product Details

SNP ID
rs6120456
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.20:31487475 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCAATCCTGTCCTCTCTGATTCTA[A/G]GTGTCTGTGAATAAGGAATGGAAAG
Phenotype
MIM: 610388
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
LINC00028 PubMed Links

Gene Details

Gene
LINC00028
Gene Name
long intergenic non-protein coding RNA 28
There are no transcripts associated with this gene.

Gene
REM1
Gene Name
RRAD and GEM like GTPase 1
There are no transcripts associated with this gene.

View Full Product Details