Product Details

SNP ID
rs8110995
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:40357935 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTATTATTAATCTCCACATTGTTCA[A/G]AGAATGAAAGTGTGGCTTAGAGAAG
Phenotype
MIM: 615698
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C19orf47 PubMed Links

Gene Details

Gene
C19orf47
Gene Name
chromosome 19 open reading frame 47
There are no transcripts associated with this gene.

Gene
PLD3
Gene Name
phospholipase D family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031696.3 Intron NP_001026866.1
NM_001291311.1 Intron NP_001278240.1
NM_012268.3 Intron NP_036400.2
XM_005258704.1 Intron XP_005258761.1
XM_005258707.4 Intron XP_005258764.1
XM_005258708.3 Intron XP_005258765.1
XM_005258709.4 Intron XP_005258766.1
XM_005258710.4 Intron XP_005258767.1
XM_006723122.1 Intron XP_006723185.1
XM_011526692.1 Intron XP_011524994.1
XM_011526693.1 Intron XP_011524995.1
XM_017026546.1 Intron XP_016882035.1
XM_017026547.1 Intron XP_016882036.1
XM_017026548.1 Intron XP_016882037.1
XM_017026549.1 Intron XP_016882038.1

View Full Product Details