Product Details

SNP ID
rs5962788
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:107127412 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACTCTATAGGCCTAACTAAATGGCT[A/G]TTGGTTAAGGTGGTATATTTGTGTA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NUP62CL PubMed Links

Gene Details

Gene
NUP62CL
Gene Name
nucleoporin 62 C-terminal like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017681.2 Intron NP_060151.2
Gene
RBM41
Gene Name
RNA binding motif protein 41
There are no transcripts associated with this gene.

View Full Product Details