Product Details

SNP ID
rs6611341
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:47174948 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCCGTCCTCTCCCCTCGGGTCCC[A/G]CCCCCGGGAACGCCGTGTGTCCAAA
Phenotype
MIM: 300080
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RBM10 PubMed Links

Gene Details

Gene
RBM10
Gene Name
RNA binding motif protein 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204466.1 Intron NP_001191395.1
NM_001204467.1 Intron NP_001191396.1
NM_001204468.1 Intron NP_001191397.1
NM_005676.4 Intron NP_005667.2
NM_152856.2 Intron NP_690595.1
XM_005272677.4 Intron XP_005272734.1
XM_005272678.4 Intron XP_005272735.1
XM_005272679.4 Intron XP_005272736.1
XM_006724563.1 Intron XP_006724626.1
XM_011543989.2 Intron XP_011542291.1
XM_017029884.1 Intron XP_016885373.1
XM_017029885.1 Intron XP_016885374.1

View Full Product Details