Product Details

SNP ID
rs9701109
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:172533319 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGCCGCTCAGCCAGCGCCATAGCC[C/T]TTAGGACTATCGGTCACATTCTCGC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SUCO PubMed Links

Gene Details

Gene
SUCO
Gene Name
SUN domain containing ossification factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282750.1 283 UTR 5 NP_001269679.1
NM_001282751.1 283 UTR 5 NP_001269680.1
NM_014283.4 283 UTR 5 NP_055098.1
NM_016227.3 283 Missense Mutation CTT,TTT L157F NP_057311.3
XM_006711374.2 283 UTR 5 XP_006711437.1
XM_006711375.2 283 UTR 5 XP_006711438.1
XM_006711376.1 283 UTR 5 XP_006711439.1

View Full Product Details