Product Details

SNP ID
rs10413640
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:48296734 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGACGGACAGACACTGACCAGGAAG[C/T]CCAGAGAACAGGAGATCAGGAGTCA
Phenotype
MIM: 615038 MIM: 603996
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCDC114 PubMed Links

Gene Details

Gene
CCDC114
Gene Name
coiled-coil domain containing 114
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144577.3 1793 UTR 3 NP_653178.3
XM_005259413.3 1793 UTR 3 XP_005259470.1
XM_005259414.3 1793 UTR 3 XP_005259471.1
XM_005259415.3 1793 UTR 3 XP_005259472.1
XM_005259416.4 1793 UTR 3 XP_005259473.1
XM_011527515.2 1793 UTR 3 XP_011525817.1
XM_011527516.2 1793 UTR 3 XP_011525818.1
XM_011527517.1 1793 Intron XP_011525819.1
XM_017027483.1 1793 UTR 3 XP_016882972.1
Gene
ZNF114
Gene Name
zinc finger protein 114
There are no transcripts associated with this gene.

View Full Product Details